Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody Type:Secondary
- Antigen Name:Immunoglobulin G
- Antigen Symbol:IgG
- Clonality:Polyclonal
- Conjugation:PerCP (Peridinin-Chlorophyll Protein Complex)
- Flow Cytometry:Yes
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Size:0.5 mL
- Form:Freeze-dried powder
- Antibody Fragment:F(ab')2
- Storage Buffer:0.01M Sodium Phosphate, 0.25M NaCl, pH 7.6, 15mg/mL BSA, 0.05% Azide
- Storage Temperature:+4 °C
- Concentration:0.5 mg/mL
- Shipping Temperature:ambient
- Purification:Chromatographically purified
- Cat. No.:102643-614
- Packaging:Glass ampoule
- Supplier no.:005-120-006
ChromPure is stands for highly purified proteins from the serum of non-immunized animals.
Type:
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: