Order Entry
Puerto Rico
ContactUsLinkComponent
Human Native alpha-2-Glycoprotein (from Plasma)
Human Native alpha-2-Glycoprotein (from Plasma)
Catalog # 10835-658
Supplier:  Avantor
CAS Number:  
undefined
Human Native alpha-2-Glycoprotein (from Plasma)
Catalog # 10835-658
Supplier:  Avantor
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/Peptide Type:
    Native
  • Source:
    Plasma
  • Species:
    Human
  • Size:
    1 mg
  • Protein Synonyms:
    Alpha-2-Z-globulin
  • Protein/Peptide Name:
    alpha-2-Glycoprotein
  • Purity:
    >90%
  • Molecular Weight:
    49.0 kDa
  • Formulation:
    Lyophilized from 20 mM Tris-HCl, pH 7.5, with 2 mM EDTA.
  • Cat. No.:
    10835-658
  • Supplier no.:
    7548-1

Specifications

About this item

≥9% pure native alpha 2 HS glycoprotein isolated from human plasma

Found in human serum at 6 mg per 1 ml. This protein is a negative acute-phase reactant. Clinically, concentrations are reduced in cancer patients. AHSG levels are positively correlated with gestational diabetes and negatively correlated with neonatal skeletal development. Studies show AHSG to be present at high concentrations in the peritoneal fluids of patients with endometriosis.