ContactUsLinkComponent
AIM™ RPR Test, Germaine Laboratories
AIM™ RPR Test, Germaine Laboratories
Catalog # 10024-416
CAS Number:  
undefined
AIM™ RPR Test, Germaine Laboratories
Catalog # 10024-416
Supplier Number:  36500
CAS Number:  
Specifications
  • Description:
    AIM™ RPR Test
  • Cat. No.:
    10024-416
  • Contents:
    RPR Antigen, Reactive Control, Weak Reactive Control, Non-Reactive Control, Dispenstir, Test Cards, Dispensing Bottle, and Dispensing Needle
  • Supplier no.:
    36500
  • No. of tests:
    500
Specifications
About this item
This microscopic non-treponemal carbon flocculation test is designed for the qualitative and quantitative detection of reagin antibodies in serum or plasma
This test is recommended for professional use, and aids in the detection of syphilis.