ContactUsLinkComponent
AIM™ CRP Test, Germaine Laboratories
  10024-410
 :  
undefined
AIM™ CRP Test, Germaine Laboratories
  10024-410
 :  32100
 :  
  • Description:
    AIM™ CRP Test
  • Cat. No.:
    10024-410
  • Contents:
    Glass Slide, Disposable Pipettes/Mixers, Positive & Negative Control, and Glycine/Saline Buffer (20x)
  • Supplier no.:
    32100
  • No. of tests:
    100
This latex agglutination test is designed for the qualitative and semi-quantitative determination of C-Reactive Protein in non-diluted serum
This test is recommended for professional use.