ContactUsLinkComponent
StrepAim® Rapid Dipstick Test, Germaine Laboratories
StrepAim® Rapid Dipstick Test, Germaine Laboratories
  10024-404
 :  
undefined
StrepAim® Rapid Dipstick Test, Germaine Laboratories
  10024-404
 :  73040
 :  
  • Description:
    StrepAim® Rapid Dipstick Test
  • Cat. No.:
    10024-404
  • Contents:
    Dipsticks, Disposable Tests Tubes, Throat Swabs, Reagents A & B, Positive & Negative Controls, and Comparison Chart
  • Supplier no.:
    73040
  • No. of tests:
    40
Dipstick method for rapid qualitative detection of Group A Strep
Test directly from throat swabs. Results are provided in 10 minutes. Store at room temperature.
 : CLIA waived.