ContactUsLinkComponent
Epredia™ Blue Nylon Biopsy Bags, Epredia
Epredia™ Blue Nylon Biopsy Bags, Epredia
  89212-290
 :  Epredia
undefined
Epredia™ Blue Nylon Biopsy Bags, Epredia
  89212-290
 :  Epredia
 :  6774015
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
  • Color:
    Blue
  • Description:
    Small
  • Size:
    Small
  • Cat. No.:
    89212-290
  • Supplier no.:
    6774015
  • Quantity per box/pack/carton:
    500 PK
Ensure unrestricted fluid movement around tissue with Epredia™ Blue Nylon biopsy bags.
  • Reduce risk of small specimen loss
  • Reduce reagent carryover and pressure artifact often found with sponges
  • Remove specimen rapidly with convenient peel-open bag