Order Entry
Puerto Rico
ContactUsLinkComponent
Advansta Buffer Pouches, Tris-Glycine-SDS
Advansta Buffer Pouches, Tris-Glycine-SDS
Catalog # 490005-040
Supplier:  Advansta
CAS Number:  
undefined
Advansta Buffer Pouches, Tris-Glycine-SDS
Catalog # 490005-040
Supplier:  Advansta
Supplier Number:  R-01036-020
CAS Number:  

Specifications

  • Description:
    Buffer SDS Tris-Glycine PAGE Pouches
  • Cat. No.:
    490005-040
  • Supplier no.:
    R-01036-020

Specifications

About this item

Convenient, pure, fast buffer preparation

  • Quality- Molecular biology-grade buffers of highest purity
  • Convenient- Pre-measured powder in sealed pouches
  • Fast- Dissolve and use right away
  • Standardized- Guaranteed reproducible results for your electrophoresis gels and Western blots

Advansta's Avant buffer pouches are perfect for standardizing your electrophoresis and Western blotting applications. Dissolve a pouch of pre-messured molecular biology-grade chemicals in 500 mL of deionized water to obtain a ready-to-use 1X buffer without any further preparation. Save precious time in the laboratory and generate reproducible results with Avant buffers.