Order Entry
Puerto Rico
ContactUsLinkComponent
JAW™ Ready-to-Use Dry Buffer Packs, G-Biosciences
JAW™ Ready-to-Use Dry Buffer Packs, G-Biosciences
Catalog # 78000-002
Supplier:  G-Biosciences
CAS Number:  
JAW™ Ready-to-Use Dry Buffer Packs, G-Biosciences
Catalog # 78000-002
Supplier:  G-Biosciences
Supplier Number:  786-291
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Concentration:
    0.2 M
  • Description:
    JAW™ Carbonate-Bicarbonate Buffer Packs [1X] (1 L/pack)
  • Size:
    20 packs
  • Cat. No.:
    78000-002
  • pH:
    9.4
  • Supplier no.:
    786-291

Specifications

About this item

Dry Powder Buffer Packs are easy-to-use, pre-mixed buffer powders that simply require adding water to generate for use.

G-Biosciences' JAW™ Carbonate-Bicarbonate Buffer packs each make one liter of: 0.2M carbonate-bicarbonate buffer (at a pH of 9.4).