ContactUsLinkComponent
Mayo Gallstone Scoop Double End, OR Grade, Sklar®
Mayo Gallstone Scoop Double End, OR Grade, Sklar®
Catalog # 10195-476
Supplier:  Sklar
undefined
Mayo Gallstone Scoop Double End, OR Grade, Sklar®
Catalog # 10195-476
Supplier:  Sklar
Supplier Number:  32-1040
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Mayo Gallstone Scoop Double End
  • Material:
    Silver Plated
  • Cat. No.:
    10195-476
  • Overall length:
    10¾"
  • Supplier no.:
    32-1040
Specifications
About this item
Sklar's® Mayo Gallstone Scoop is a rigid, double-ended gall bladder instrument with spoon shaped tips used for removing gallstones from the gall bladder
This product is slightly curved at each end.