Specifications
- Description:Mayo Duct Scoop, Small
- Length:10½"
- Material:Silver Plated
- Cat. No.:10195-472
- Supplier no.:32-1014
Specifications
About this item
The Sklar® Mayo Duct Scoop is a malleable, silver-plated instrument used for removing gallstones from the gall bladder or common bile duct
They are available with three scoop sizes to accommodate the specific procedure.