Order Entry
Puerto Rico
ContactUsLinkComponent
 
AIX1000™ Fully Automated RPR Syphilis Testing System, Gold Standard Diagnostics
 
undefined
AIX1000™ Fully Automated RPR Syphilis Testing System, Gold Standard Diagnostics

 

  • Catalog No:
  • 10853-848
  • 10853-092
  • Sample capacity:
  • 192 Patient Sample Positions
  • 192 Patient Sample Positions
  • Electrical:
  • 120–265V, 50/60Hz, 120W max.
  • 120–265V, 50/60Hz, 120W max.
  • W×D×H:
  • 64×57×45 cm (25.2×22.4×17.7")
  • 64×57×45 cm (25.2×22.4×17.7")
  • Weight:
  • 28 kg (61.7 lbs.)
  • 28 kg (61.7 lbs.)