Order Entry
Puerto Rico
ContactUsLinkComponent
TRIS HCl (tris(hydroxymethyl)aminomethane hydrochloride) ≥99.5%
TRIS HCl (tris(hydroxymethyl)aminomethane hydrochloride) ≥99.5%
Catalog # IB70162
Supplier:  IBI Scientific
CAS Number:  
undefined
TRIS HCl (tris(hydroxymethyl)aminomethane hydrochloride) ≥99.5%
Catalog # IB70162
Supplier:  IBI Scientific
Supplier Number:  IB70162
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    500 g
  • Cat. No.:
    IB70162
  • Supplier no.:
    70162

Specifications

About this item

 :
 :157.6 g/mol
Merck Index:12,09902
Melting Pt:150…152 °C
Storage Temperature:Ambient
EINECS:214-684-5
CAS Number:1185-53-1
Formula:NH₂C(CH₂OH)₃·HCl
MDL Number:MFCD00012590