ContactUsLinkComponent
Human Bladder Fibroblasts, Lifeline Cell Technology
Human Bladder Fibroblasts, Lifeline Cell Technology
Catalog # 102970-932
CAS Number:  
undefined
Human Bladder Fibroblasts, Lifeline Cell Technology
Catalog # 102970-932
Supplier Number:  FC-0050
CAS Number:  
Specifications
  • Cell Format:
    Cryopreserved
  • Description:
    Human Bladder Fibroblasts
  • Size:
    500,000 cells per vial
  • Cat. No.:
    102970-932
  • Supplier no.:
    FC-0050
Specifications
About this item
Lifeline® Normal Human Bladder Fibroblasts are quality tested and demonstrate optimal growth.
  • Tested for morphology
  • Can be grown in serum-free, phenol red-free, and antimicrobial-free conditions
  • Quality tested for optimal growth
Lifeline® normal Human Bladder Fibroblasts are quality tested in FibroLife® S2 or FibroLife® Xeno-Free Medium and demonstrate optimal growth at rates equal to or greater than classical serum-supplemented media
Bladder fibroblasts can be grown in serum-free, phenol red-free, and antimicrobial-free conditions when cultured in FibroLife® Serum-Free Medium.
Lifeline® bladder fibroblasts are tested for proper growth and morphology over a period of at least 15 population doublings.