Bronchial/Tracheal Epithelial Cells, Lifeline Cell Technology
Catalog # 102970-680
Supplier: Lifeline Cell Technology
CAS Number:
Specifications
- Cell Format:Cryopreserved
- Description:Human Bronchial/Tracheal Epithelial Cells, Primary
- Size:500,000 cells per vial
- Cat. No.:102970-680
- Supplier no.:FC-0035
Specifications
About this item
Lifeline® Normal Human Bronchial/Tracheal Epithelial Cells ensure the highest viability and plating efficiency.
- Quality tested to ensure optimal growth
- Cultured without retinoic acid
- Cryopreserved as Primary cells
- No exposure to antimicrobials or phenol red
Lifeline® normal Human Bronchial/Tracheal Epithelial Cells are cultured without retinoic acid and cryopreserved as Primary cells to ensure the highest viability and plating efficiency
Bronchial/Tracheal Epithelial Cells are not exposed to antimicrobials or phenol red when cultured in Lifeline® BronchiaLife™ medium.
Our Bronchial/Tracheal Epithelial Cells are quality tested in BronchiaLife™ Medium to ensure optimal growth over a period of at least 15 population doublings at rates equal to or greater than other commercially available media.