rh IGF-1 LifeFactor, Lifeline Cell Technology
Catalog # 102970-734
Supplier: Lifeline Cell Technology
CAS Number:
Specifications
- Description:rh IGF-1 LifeFactor
- Size:0.5 mL
- Cat. No.:102970-734
- Supplier no.:LS-1014
Specifications
About this item
Recombinant human insulin-like growth factor is used with basal medium and human cells.
- Used with human endothelial cells
- Used with microvascular endothelial cells
- Used with basal medium
Recombinant human insulin-like growth factor (rh IGF-1) may be used with Lifeline® VascuLife® basal medium and human endothelial cells, including microvascular endothelial.