Orders LinkContactUsLinkComponent
NEFA Linearity Set Solution, Wako
NEFA Linearity Set Solution, Wako
NEFA Linearity Set Solution, Wako
NEFA Linearity Set Solution, Wako
The NEFA Linearity Set is a 2.5 mEq/L solution used to verify the measurable range of the NEFA HR(2) assay.
This product is only used with Wako’s NEFA HR(2) series of reagents.
  • Catalog No:
  • 80076-166
  • 97000-010
  • 97000-014
  • 97000-012
  • 97000-016
  • Linearity:
  • 0.01 - 4.00 mEq/L
  • 0.01 - 4.00 mEq/L
  • 0.01 - 4.00 mEq/L
  • 0.01 - 4.00 mEq/L
  • Method:
  • Enzymatic
  • Enzymatic
  • Enzymatic
  • Enzymatic
  • Stability:
  • 30 days after reconstitution
Product Family Options
  • Description
    Size
    Storage temperature
  • NEFA Standard Solution
    4×10 ml
    +2…+10 °C
    Pack of 4
    Pack of 4 - $252.94
    $252.94
    Pack of 4
       $252.94
     : 276-76491
    Specifications
    Cat. No.
    80076-166
    Linearity
    Method
    Stability
  • NEFA-HR(2) Color A
    4×50 ml
    +2…+10 °C
    Each
    Each - $828.29
    $828.29
    Each
       $828.29
     : 999-34691
    Specifications
    Cat. No.
    97000-010
    Linearity
    0.01 - 4.00 mEq/L
    Method
    Enzymatic
    Stability
    30 days after reconstitution
  • NEFA-HR(2) Color B
    4×25 ml
    +2…+10 °C
    Each
    Each - $673.98
    $673.98
    Each
       $673.98
     : 991-34891
    Specifications
    Cat. No.
    97000-014
    Linearity
    0.01 - 4.00 mEq/L
    Method
    Enzymatic
    Stability
  • NEFA-HR(2) Solvent A
    4×50 ml
    +2…+10 °C
    Each
    Each - $133.66
    $133.66
    Each
       $133.66
     : 995-34791
    Specifications
    Cat. No.
    97000-012
    Linearity
    0.01 - 4.00 mEq/L
    Method
    Enzymatic
    Stability
  • NEFA-HR(2) Solvent B
    4×25 ml
    +2…+10 °C
    Each
    Each - $222.92
    $222.92
    Each
       $222.92
     : 993-35191
    Specifications
    Cat. No.
    97000-016
    Linearity
    0.01 - 4.00 mEq/L
    Method
    Enzymatic
    Stability
Some Products May Appear Restricted