ContactUsLinkComponent
Human Bladder Smooth Cells, Lifeline Cell Technology
Human Bladder Smooth Cells, Lifeline Cell Technology
Catalog # 102970-692
CAS Number:  
undefined
Human Bladder Smooth Cells, Lifeline Cell Technology
Catalog # 102970-692
Supplier Number:  FC-0043
CAS Number:  
Specifications
  • Cell Format:
    Cryopreserved
  • Description:
    Human Bladder Smooth Muscle Cells
  • Size:
    500,000 cells per vial
  • Cat. No.:
    102970-692
  • Supplier no.:
    FC-0043
Specifications
About this item
Lifeline® Normal Human Bladder Smooth Muscle Cells are quality tested and demonstrate optimal low serum growth.
  • Quality tested in VascuLife® SMC Medium
  • Demonstrate optimal low serum growth
  • No exposure to antimicrobials or phenol red
Lifeline® Normal Human Bladder Smooth Muscle Cells are quality tested in VascuLife® SMC Medium and demonstrate optimal low serum growth over a period of at least 15 population doublings at rates equal to or greater than other serum-supplemented media.
Lifeline® Bladder Smooth Muscle Cells are not exposed to antimicrobials or phenol red when cultured in VascuLife® SMC Medium.