Neonatal Microvascular Endothelial Cells, Lifeline Cell Technology
Catalog # 102970-690
Supplier: Lifeline Cell Technology
CAS Number:
Specifications
- Cell Format:Cryopreserved
- Description:Human Dermal Microvascular Endothelial Cells, Neonatal
- Size:500,000 cells per vial
- Cat. No.:102970-690
- Supplier no.:FC-0042
Specifications
About this item
Lifeline® Normal Human Dermal Microvascular Neonatal Endothelial Cells are cryopreserved and can be cultured with or without VEGF.
- Exposure to antimicrobials or phenol red unnecessary
- Cryopreserved after less than four weeks in culture
- Can be cultured with or without VEGF
Lifeline® Normal Human Dermal Microvascular Neonatal Endothelial Cells need not be exposed to antimicrobials or phenol red when cultured in Lifeline® VascuLife® medium.
Microvascular Endothelial Cells are cryopreserved after less than four weeks in culture. They can be cultured with or without VEGF.