Orders LinkContactUsLinkComponent
Lipid Quantification Kit, Cell Biolabs
Marketsource
Lipid Quantification Kit, Cell Biolabs
Marketsource
Lipid Quantification Kit, Cell Biolabs
Lipid Quantification Kit, Cell Biolabs
Cell Biolabs' Lipid Quantitation Kit provides a simple method for quantitation of unsaturated fatty acids from plasma, serum, or cultured cells in a standard 96-well microplate reader
The kit uses a sulfo-phospho-vanillin system in which samples are acidified and heated to solubilize and prime them for quantitation. In an acidic environment, vanillin reagent reacts with the lipids to form a colored product that is easily detected by the plate reader at 540 nm.
  • Catalog No:
  • 102965-228
  • 102965-236
  • Storage Temperature:
  • –20°C
  • 4°C
Product Family Options
  • Description
    Size
    Method
  • Lipid Quantification Kit
    100 Assays
    Colorimetric
    Each
    Each - $590.77
    $590.77
    Each
       $590.77
     : STA-613
    Marketsource
    Specifications
    Cat. No.
    102965-228
    Storage Temperature
    –20°C
    Marketsource
  • Lipid Quantification Kit
    100 Assays
    Fluorometric
    Each
    Each - $590.77
    $590.77
    Each
       $590.77
     : STA-617
    Marketsource
    Specifications
    Cat. No.
    102965-236
    Storage Temperature
    4°C
    Marketsource