ChondroLife™ Complete Medium, Lifeline Cell Technology
Catalog # 102970-778
Supplier: Lifeline Cell Technology
CAS Number:
Specifications
- Description:ChondroLife™ Chondrogenesis Complete Frozen Medium
- Size:100 ml
- Cat. No.:102970-778
- Supplier no.:LM-0022
Specifications
About this item
ChondroLife™ is a serum-free medium optimized for the differentiation of adult human mesenchymal stem cells.
- Produces chondrocytes
- Exhibits superior deposition of sulfated proteoglycan
- No antimicrobials
- No phenol red
- Fully-supplemented frozen medium
ChondroLife™ is a serum-free medium optimized for the differentiation of adult human mesenchymal stem cells into chondrocytes. ChondroLife™ produces chondrocytes that exhibit superior deposition of sulfated proteoglycans as compared with other commercially available products. ChondroLife™ contains no antimicrobials or phenol red.
ChondroLife™ is a fully-supplemented frozen medium.