Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Alizarin Red S (sodium salt)
Alizarin red S (sodium salt) 2% in aqueous solution stain
Alizarin red S (sodium salt) 2% in aqueous solution stain
102970-892
:
Lifeline Cell Technology
:
Alizarin red S (sodium salt) 2% in aqueous solution stain
102970-892
:
Lifeline Cell Technology
:
CM-0058
:
Size:
100 ml
Cat. No.:
102970-892
Supplier no.:
CM-0058
:
:
342.26 g/mol
:
GHS07
:
130-22-3
:
93982-72-0
:
C₁₄H₇NaO₇S
:
MFCD00013049