Phosphate Buffered Solution Xeno-Free, Lifeline Cell Technology
Catalog # 102970-802
Supplier: Lifeline Cell Technology
CAS Number:
Specifications
- Description:Phosphate Buffered Solution, Xeno Free
- Size:500 mL
- Cat. No.:102970-802
- Supplier no.:CM-0001
Specifications
About this item
Phosphate Buffered Saline is used to remove media serum before trypsinization.
- Maintains a constant pH
- Ion concentration and osmolarity matches that of human cells
- Contains no non-human animal-derived components
- Does not contain calcium or magnesium
Phosphate Buffered Saline (PBS) Xeno-Free maintains a constant pH, and its ion concentration and osmolarity matches that of human cells. PBS is used to remove media serum before trypsinization. Contains no non-human animal-derived components. Does not contain calcium or magnesium.