Trypsin Neutralizing Solution, Lifeline Cell Technology
Supplier: Lifeline Cell Technology
Certificates
About this item
Trypsin acts as an enzyme to help dissociate cells from their culture vessel.
- Acts as an enzyme to break down proteins
Trypsin acts as an enzyme to break down proteins to dissociate cells from the vessel in which they're being cultured. Trypsin Neutralizing Solution Xeno-Free contains no non-human animal-derived component.