ContactUsLinkComponent
AdipoLife™ DfKt™-1 Adipogenesis Kit (for Adipose MSC and Pre-Adipocytes), Lifeline Cell Technology
AdipoLife™ DfKt™-1 Adipogenesis Kit (for Adipose MSC and Pre-Adipocytes), Lifeline Cell Technology
Catalog # 102970-900
undefined
AdipoLife™ DfKt™-1 Adipogenesis Kit (for Adipose MSC and Pre-Adipocytes), Lifeline Cell Technology
Catalog # 102970-900
Supplier Number:  LL-0050
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    AdipoLife™ DfKt™-1 Adipogenesis Kit (for Adipose MSC and Pre-Adipocytes)
  • Size:
    100 mL
  • Cat. No.:
    102970-900
  • Supplier no.:
    LL-0050
Specifications
About this item
DfKt™-1 Adipogenesis Medium is optimized for the differentiation of human adipose-derived Mesenchymal Stem Cells.
  • Differentiation of human adipose-derived mesenchymal stem cells
  • Optimized for pre-adipocytes into adipocytes
  • No antimicrobials
  • No phenol red
AdipoLife™ DfKt™-1 Adipogenesis Medium is optimized for the differentiation of human adipose-derived mesenchymal stem cells and pre-adipocytes into adipocytes.
AdipoLife™ DfKt™-1 contains no antimicrobials and no phenol red.
Ordering information: Use AdipoLife™ Adipogenesis Kit 1 with Adipose-derived mesenchymal stem cells and pre-adipocytes.