Trypkit™ Large, Lifeline Cell Technology
Catalog # 102970-818
Supplier: Lifeline Cell Technology
CAS Number:
Specifications
- Description:TrypKit™ (PBS, Trypsin/EDTA), large
- Size:100 ml
- Cat. No.:102970-818
- Supplier no.:LL-0013
Specifications
About this item
Delivery: The kit includes phosphate buffered saline (PBS) (500 ml), 0.05% trypsin/0.02% EDTA (phenol red-free) (100 ml), trypsin neutralizing solution (100 ml).
Caution: TrypKit™ is for research use only. This product is not approved for human or veterinary use or for use in in vitro diagnostics or clinical procedures.