Cdc42 Activation Assays, Cell Biolabs
Catalog # 102965-110
Supplier: Cell Biolabs
CAS Number:
Specifications
- Description:Cdc42 Activation Assay
- Size:20 Assays
- Storage Temperature:–20°C
- Cat. No.:102965-110
- Method:Immunoblot
- Supplier no.:STA-402
Specifications
About this item
Cell Biolabs' Cdc42 Activation Assays use visible agarose beads to selectively precipitate the active form of Cdc42 protein
The precipitated small GTPase is then detected by Western blot using a Cdc42-specific antibody included in the kit. The assay uses a safe, non-radioactive format.