ContactUsLinkComponent
Sklar Black™ Graves Wide-View Vaginal Speculum, OR Grade, Sklar®
  10184-274
 :  Sklar
undefined
Sklar Black™ Graves Wide-View Vaginal Speculum, OR Grade, Sklar®
  10184-274
 :  Sklar
 :  90-9055
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
  • Sterility:
    Non sterile
  • Disposable/Reusable:
    Reusable
  • Latex/Latex-free:
    Latex-Free
  • Material:
    Stainless Steel with Reduced Reflectivity Polymer Coating
  • Size:
    Medium with Loop
  • Tip dimensions:
    4×11/4"
  • Curvature:
    Straight
  • Cat. No.:
    10184-274
  • Supplier no.:
    90-9055
The Sklar Black™ Graves Wide-View Vaginal Speculum is used for laser gynecological procedures
  • Industry standard
It is a double blade duckbill speculum inserted with the blades closed. The blades are then opened by an external lever and screw. The Sklar Black™ Graves Vaginal Speculum is available with a permanent smoke evacuation tube, a loop for disposable smoke evacuation tubing, or no smoke evacuation. Sklar Black™ Laser instruments feature a non-reflective black polymer coating applied over the sandblasted instrument developed specifically for laser surgeries. This safe, durable coating reduces laser reflection to less than one-tenth of a percent, ensuring a safer surgical environment for patients and surgical staff.
 : Pre-Vacuum and Gravity Cycle Steam Sterilization Validation of ANSI/AAMI ST79.