ContactUsLinkComponent
All Purpose Brush Set, Sklar®
All Purpose Brush Set, Sklar®
Catalog # 10161-766
Supplier:  Sklar
undefined
All Purpose Brush Set, Sklar®
Catalog # 10161-766
Supplier:  Sklar
Supplier Number:  10-1434
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    All Purpose Brush Set, Stainless Steel/Plastic/Nylon
  • Cat. No.:
    10161-766
  • Supplier no.:
    10-1434
Specifications
About this item
All Purpose Brush Set Includes # 10-1650 Brush Stainless Steel Bristle 3/pk, #10-1651 Brush Nylon Bristle 3/pk, #10-1420 Large Instrument Cleaning Brush 3/pk, #10-1418 Surface Cleaning Brush 3/pk - Set
Sklar's® All Purpose Brush Set contains 4 varieties of instrument cleaning brushes that can be used on multiple surfaces.