ContactUsLinkComponent
Garrett Vascular Dilators Set, OR Grade, Sklar®
  10197-246
 :  Sklar
undefined
Garrett Vascular Dilators Set, OR Grade, Sklar®
  10197-246
 :  Sklar
 :  52-7000
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
  • Description:
    Garrett Vascular Dilators Set of 9, Short
  • Length:
    5½"
  • Material:
    Stainless steel
  • Cat. No.:
    10197-246
  • Supplier no.:
    52-7000
Sklar's® Garrett Vascular Dilator is designed to maneuver through vessels during cardiothoracic surgical procedures.
It is important for surgeons to be able to measure the diameter of vessels and these vascular dilators are suited to perform that specific task.
This product is a set of 9 short (5-1/2 inch overall length) dilators with 1mm, 1.5mm, 2mm, 2.5mm, 3mm, 3.5mm, 4mm, 4.5mm, and 5mm tips.