QuickTiter™ MuLV Core Antigen ELISA Kit, Cell Biolabs
Catalog # 102966-344
Supplier: Cell Biolabs
CAS Number:
Specifications
- Conjugate ELISA:HRP
- Description:MuLV Core Antigen ELISA Kit
- Size:96 Assays
- Detection Method:Colorimetric
- Storage Temperature:4°C and –20°C
- Sensitivity:300 pg/mL
- Cat. No.:102966-344
- Supplier no.:VPK-156
Specifications
About this item
Cell Biolabs’ QuickTiter™ MuLV Core Antigen ELISA Kit is an enzyme immunoassay developed for detection and quantitation of the MuLV core antigen protein (gag p30)
The kit has a detection sensitivity limit of 300 pg/mL MuLV p30. Each kit provides sufficient reagents to perform up to 96 assays including standard curve and MuLV samples.