QuickTiter™ HCV Core Antigen ELISA Kit, Cell Biolabs
102966-342
:
- Conjugate ELISA:HRP
- Description:HCV Core Antigen ELISA Kit
- Size:96 Assays
- Detection Method:Colorimetric
- Storage Temperature:4°C and –20°C
- Sensitivity:1 ng/mL
- Cat. No.:102966-342
- Supplier no.:VPK-151
Cell Biolabs’ QuickTiter™ HCV Core Antigen ELISA Kit is an enzyme immunoassay developed for detection and quantitation of the HCV core protein
The kit has detection sensitivity limit of 1 ng /mL HCVcAg. Each kit provides sufficient reagents to perform up to 96 assays including standard curve and HCV samples.