QuickTiter™ HBV Core Antigen ELISA Kit, Cell Biolabs
Cell Biolabs’ QuickTiter™ HBV Core Antigen ELISA Kit is an enzyme immunoassay developed for detection and quantitation of the HBV core protein
The kit has detection sensitivity limit of 1 ng/mL HBcAg. Each kit provides sufficient reagents to perform up to 96 assays including standard curve and HBV samples.
- Conjugate ELISA:HRP
- Detection Method:Colorimetric
- Storage Temperature:4°C and –20°C
- Sensitivity:1 ng/mL
Frequently Bought Together