QuickTiter™ Lentivirus Quantitation Kit, Cell Biolabs
Catalog # 102965-356
Supplier: Cell Biolabs
CAS Number:
Specifications
- Description:Lentivirus Quantitation Kit
- Size:20 Assays
- Storage Conditions:4°C
- Cat. No.:102965-356
- Supplier no.:VPK-112
Specifications
About this item
QuickTiter™ Lentivirus Quantitation Kit provides a quick method for measuring the viral nucleic acid content of your lentivirus prep
This assay may be performed either before or after purification of your virus. The kit uses an ultra-fast 45-60 minute procedure to measure the viral nucleic acid content of purified lentivirus or unpurified supernatant. The limit of detection is 10^10 viral particles/mL from 100 µL of lentiviral supernatant.