Order Entry
Puerto Rico
ContactUsLinkComponent
QuickTiter™ Adenovirus Quantitation Kit, Cell Biolabs
QuickTiter™ Adenovirus Quantitation Kit, Cell Biolabs
Catalog # 102965-342
Supplier:  Cell Biolabs
CAS Number:  
undefined
QuickTiter™ Adenovirus Quantitation Kit, Cell Biolabs
Catalog # 102965-342
Supplier:  Cell Biolabs
Supplier Number:  VPK-106
CAS Number:  

Specifications

  • Description:
    Adenovirus Quantitation Kit
  • Size:
    20 Assays
  • Storage Temperature:
    4°C
  • Sensitivity:
    10^10 VP/mL
  • Cat. No.:
    102965-342
  • Supplier no.:
    VPK-106

Specifications

About this item

QuickTiter™ Adenovirus Quantitation Kit provides a quick method for measuring the viral nucleic acid content of your adenovirus prep

This assay may be performed either before or after purification of your virus. The kit uses an ultra-fast 45-60 minute procedure to measure the viral nucleic acid content of purified adenovirus or unpurified supernatant. The limit of detection is 10^10 viral particles/mL from 100 µL of adenoviral supernatant.