To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Assay Type (ELISA with LOV):Sandwich
- Conjugate ELISA:HRP
- Host:
- Primary antibody reactivity:
- Description:Protein Carbamylation ELISA Kit
- Size:96 Assays
- Environmentally Preferable:
- Detection Method:Colorimetric
- Storage Temperature:4°C and –20°C
- Sensitivity:1.5 ng/mL
- Cat. No.:102965-328
- Supplier no.:STA-877
Specifications
About this item
The OxiSelect™ Protein Carbamylation Sandwich ELISA Kitis an enzyme immunoassay developed for rapid detection and quantitation of protein carbamylation
CBL-BSA standards or protein samples are added to an Anti-CBL Antibody Coated Plate for 2 hours at 37ºC. The CBL protein adducts present in the sample or standard are probed with an anti-CBL antibody, followed by an HRP conjugated secondary antibody. The protein CBL adduct content in an unknown sample is determined by comparing against a standard curve prepared from CBL-BSA standards. This kit can detect as little as 1.5 ng/mL of CBL-BSA.