OxiSelect™ HNE Adduct Competitive ELISA Kit, Cell Biolabs
Supplier: Cell Biolabs
Certificates
About this item
OxiSelect™ HNE Adduct Competitive ELISA Kit provides a sensitive, specific method for detection of malondialdehyde adducts to proteins
4-HNE, or 4-hydroxynonenal, is a widely accepted marker for oxidative stress and is a by-product of lipid peroxidation.
Specifications
- Assay Type (ELISA with LOV):Competitive
- Conjugate ELISA:HRP
- Detection Method:Colorimetric
- Storage Temperature:4°C and –20°C
- Sensitivity:2 µg/ml