About this item
OxiSelect™ AGE Competitive ELISA kit is designed for rapid detection and quantitation of advanced glycation end product protein adducts formed during the Maillard reaction, where reducing carbohydrates react with lysine side chains and N-terminal amino groups of various macromolecules, particularly proteins
The quantity of AGE adduct in protein samples is determined by comparing its OD with that of a known AGE-BSA standard curve. This kit detects AGE adduct levels as low as 0.5 µg/mL.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Assay Type (ELISA with LOV):Competitive
- Conjugate ELISA:HRP
- Detection Method:Colorimetric
- Storage Temperature:4°C and –20°C
- Sensitivity:500 ng/mL