ContactUsLinkComponent
Poly (D,L-lactide-co-glycolide), 50:50, IV 1.0 dl/g, Polysciences
  87001-914
Small Business Enterprise
 :  
undefined
Poly (D,L-lactide-co-glycolide), 50:50, IV 1.0 dl/g, Polysciences
  87001-914
 :  23987-5
Small Business Enterprise
 :  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
  • Description:
    Poly (D,L-lactide-co-glycolide), 50:50, IV 1.0 dl/g
  • Size:
    5 g
  • Cat. No.:
    87001-914
  • Supplier no.:
    23987-5
Poly (DL-lactide-co-glycolide) (PLGA) is a well-established biodegradable polymer used in medical devices and drug delivery applications
In comparison to Poly (L-lactide) (PLLA), PLGA offers control over degredation rate and reduced glass transition temperatures (by increasing glycolide content). The control of these properties makes PLGA especially well suited for applications where medical devices are only needed for weeks (e.g. drug delivery).