Order Entry
Puerto Rico
ContactUsLinkComponent
QuickTiter™ Hepatitis B Surface Antigen (HBsAg) ELISA Kit, Cell Biolabs
QuickTiter™ Hepatitis B Surface Antigen (HBsAg) ELISA Kit, Cell Biolabs
Supplier:  Cell Biolabs
Certificates
undefined
QuickTiter™ Hepatitis B Surface Antigen (HBsAg) ELISA Kit, Cell Biolabs
Supplier:  Cell Biolabs

Specifications

  • Assay Type (ELISA with LOV):
    Sandwich
  • Conjugate ELISA:
    HRP
  • Detection Method:
    Colorimetric
  • Storage Temperature:
    4°C and –20°C
  • Sensitivity:
    1 ng/mL

Specifications

Related Information