QuickTiter™ Hepatitis B 'e' Antigen (HBeAg) ELISA Kit, Cell Biolabs
Supplier: Cell Biolabs
Certificates
About this item
QuickTiter™ HBeAg ELISA is a sandwich ELISA that is designed for detection and quantitation of Hepatitis B 'e' antigen in purified virus or unpurified viral supernatant
This assay uses an anti-HBeAg antibody coated plate that binds to any HBeAg present in the sample. A FITC-conjugated anti-HBeAg antibody is then added, which binds to HBeAg sample-antibody complex. This is detected by the addition of an HRP-conjugated anti-FITC antibody. HBeAg levels in unknown samples are calculated based on the HBeAg standard curve. This kit detects as little as 150 pg/mL of HBeAg.
Specifications
- Assay Type (ELISA with LOV):Sandwich
- Conjugate ELISA:HRP
- Detection Method:Colorimetric
- Storage Temperature:4°C and –20°C
- Sensitivity:150 pg/mL
Specifications
Frequently Bought Together