Order Entry
Puerto Rico
ContactUsLinkComponent
 
Corning® Polypropylene Cap with 2 Stainless Steel Tubing Ports, Corning
  89089-806
 :  Corning
Product available on GSA Advantage®
undefined
Corning® Polypropylene Cap with 2 Stainless Steel Tubing Ports, Corning
  89089-806
 :  Corning
 :  401654
Product available on GSA Advantage®

 

  • Description:
    45 mm Cap with 2 Stainless Steel Tubing Ports
  • Material:
    PP
  • Cat. No.:
    89089-806
  • Thread size:
    45 GL
  • Supplier no.:
    401654

 

 

Autoclavable polypropylene caps feature two 15.2 cm (6") stainless steel tubes

Caps fit Corning glass and plastic vessels with GL45 mm threads. Designed for bioprocess and scale-up applications.