Corning® PBT GL 32 Caps, Corning
Catalog # 89089-690
Supplier: Corning
Product available on GSA Advantage®
Specifications
- Color:Red
- Material:PBT
- Environmentally Preferable:Low Manufacturing Impact
- Environmental Certification:I-RECs, ISO 14001 - Monterrey
- Cat. No.:89089-690
- Thread finish:GL 32
- Supplier no.:1395-32HTC
Specifications
About this item
The red Corning® polybutylene terephthalate (PBT) high temperature caps have a GL32 thread, and can be dry heat sterilized at up to 180 °C
Caps have a Polytetrafluorethylene (PTFE) liner for efficient sealing.