Serum Triglyceride Quantification Kits, Cell Biolabs
Cell Biolabs’ Serum Triglyceride Quantification Kits measure triglyceride concentrations in serum, plasma, and lysates by a coupled enzymatic reaction system. First, lipase hydrolyzes the triglyceride ester bond, yielding glycerol. The free glycerol is then phosphorylated and oxidized, producing hydrogen peroxide which reacts with the kit’s probe
Kits are available with either colorimetric or fluorometric detection in a 96-well plate reader.
Cell Biolabs’ Serum Triglyceride Quantification Kits measure triglyceride concentrations in serum, plasma, and lysates by a coupled enzymatic reaction system. First, lipase hydrolyzes the triglyceride ester bond, yielding glycerol. The free glycerol is then phosphorylated and oxidized, producing hydrogen peroxide which reacts with the kit’s probe
Kits are available with either colorimetric or fluorometric detection in a 96-well plate reader.
- Catalog No:
- 102965-078
- 102965-080
- Sensitivity:
- 1 mg/dL (colorimetric), 0.2 mg/dL (fluorometric)
- 1 mg/dL (colorimetric), 0.2 mg/dL (fluorometric)
- Storage Temperature:
- –20°C
- –20°C
Product Family Options
- DescriptionSizeMethod
- : STA-396Marketsource
Specifications
Cat. No.102965-078Sensitivity1 mg/dL (colorimetric), 0.2 mg/dL (fluorometric)Storage Temperature–20°CMarketsource - : STA-397Marketsource
Specifications
Cat. No.102965-080Sensitivity1 mg/dL (colorimetric), 0.2 mg/dL (fluorometric)Storage Temperature–20°CMarketsource



