OxiSelect™ UV-Induced DNA Damage ELISA Combo Kit (CPD/6-4PP Quantitation), Cell Biolabs
Catalog # 102964-914
Supplier: Cell Biolabs
CAS Number:
Specifications
- Conjugate ELISA:HRP
- Description:CPD and 6-4PP Quantitation ELISA Combo Kit
- Size:96 Assays
- Detection Method:Colorimetric
- Storage Temperature:4°C and –20°C
- Cat. No.:102964-914
- Supplier no.:STA-322-C
Specifications
About this item
OxiSelect™ UV-Induced DNA Damage ELISA Combo Kit measures the formation of both CPD and 6-4PP structures in DNA isolated from cells or tissues
Standards or unknown DNA samples are first heat denatured before adsorbed onto a 96-well DNA binding plate. The sample or standard are then probed with an anti-6-4PP or CPD antibody, followed by an HRP conjugated secondary antibody. CPD and 6-4PP structures are assayed on the same plate but in separate wells.