Order Entry
Puerto Rico
ContactUsLinkComponent
Lentiviral Control Vectors, Cell Biolabs
Lentiviral Control Vectors, Cell Biolabs
Supplier:  Cell Biolabs
Certificates
undefined
Lentiviral Control Vectors, Cell Biolabs
Supplier:  Cell Biolabs

Specifications

  • Format:
    Bacterial Glycerol Stock
  • Storage Temperature:
    –80°C

Specifications

Related Information