StemTAG™ Stem Cell Colony Formation Assay, Cell Biolabs
StemTAG™ 96-Well Stem Cell Colony Formation Assay provides a high-throughput method to quantify ES cells in just 7-10 days, and no manual cell counting is required
Once colonies are formed, they may be analyzed in three different ways: (a) lyse cells, then quantify in a fluorescence plate reader using dye included in the kit; (b) lyse cells, then quantify quantify alkaline phosphatase activity using reagents provided; (c) recover colonies from matrix for further culture or analysis.
- Storage Temperature:4°C