Order Entry
Puerto Rico
ContactUsLinkComponent
SNL Passage-Independent Feeder Cells, Cell Biolabs
SNL Passage-Independent Feeder Cells, Cell Biolabs
Catalog # 102963-872
Supplier:  Cell Biolabs
CAS Number:  
undefined
SNL Passage-Independent Feeder Cells, Cell Biolabs
Catalog # 102963-872
Supplier:  Cell Biolabs
Supplier Number:  CBA-316
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Cell Format:
    Cryopreserved
  • Description:
    SNL Feeder Cells
  • Size:
    1 Vial
  • Storage Conditions:
    –80°C or LN2
  • Cat. No.:
    102963-872
  • Supplier no.:
    CBA-316

Specifications

About this item

SNL 76/7 Feeder Cells are clonally derived from a mouse fibrobast STO cell line which has been transformed with neomycin resistance and murine LIF genes

This immortalized cell line has been shown to be useful as a feeder cell for ES cell growth as well as for culture of human and mouse iPS cells.