Anti-IFNG Mouse Monoclonal Antibody [clone: 1.1-5A9-2D05]
Catalog # 10816-218
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:interferon, gamma
- Antigen Symbol:IFNG
- Clonality:Monoclonal
- Clone:1.1-5A9-2D05
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western Blot:Yes
- Size:0.5 mg
- Form:Lyophilized
- Gene ID:P01579
- Antigen Synonyms:IFNG
- Storage Temperature:Stable at room temperature for at least one month and for greater than a year when kept at –20 °C. Reconstitute in sterile water to a concentration of 0.1 - 1.0 mg/mL. Stable for at least six weeks at 2-4 °C when reconstituted.
- Shipping Temperature:RT
- Immunogen:The monoclonal antibody was produced in BALB/c x ICR F1 mice using recombinant IFN-gamma as the immunizing antigen.
- Purification:This IgG1k antibody was purified on a Protein A affinity column.
- Cat. No.:10816-218
- Supplier no.:XP-5739-M
Specifications
About this item
ELISA: In a sandwich ELISA (assuming 100 uL/well), a concentration of 4.0-8.0 ug/mL of this antibody will detect at least 500 pg/mL of recombinant human IFN-gamma when used with our biotinylated antigen affinity purified anti-human IFN-gamma (XP-5155Bt) as the detection antibody at a concentration of approximately 2.0-4.0 ug/mL. Western Blot: To detect hIFN-gamma by Western Blot analysis this antibody can be used at a concentration of 0.20-0.40 ug/mL. Used in conjunction with compatible secondary reagents the detection limit for recombinant hIFN-gamma is 1.0-3.0 ng/lane, under non-reducing conditions.
Type: Primary
Antigen: IFNG
Clonality: Monoclonal
Clone: 1.1-5A9-2D05
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human