Anti-CCL2 Mouse Monoclonal Antibody [clone: 2.2-4H5-01A11]
Catalog # 10813-682
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:chemokine (C-C motif) ligand 2
- Antigen Symbol:CCL2
- Clonality:Monoclonal
- Clone:2.2-4H5-01A11
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western Blot:Yes
- Size:0.5 mg
- Form:Lyophilized
- Gene ID:P13500
- Antigen Synonyms:CCL2
- Storage Temperature:Stable at room temperature for at least one month and for greater than a year when kept at –20 °C. Reconstitute in sterile water to a concentration of 0.1 - 1.0 mg/mL. Stable for at least six weeks at 2-4 °C when reconstituted.
- Shipping Temperature:RT
- Immunogen:The monoclonal antibody was produced in BALB/c X ICR F1 mice using recombinant hMCP-1/MCAF as the immunizing antigen.
- Purification:This IgG1/K antibody was purified from ascites fluid on a Protein A affinity column.
- Cat. No.:10813-682
- Supplier no.:XP-5732-M
Specifications
About this item
ELISA: In a sandwich ELISA (assuming 100 uL/well), a concentration of 2.0-4.0 ug/mL of this antibody will detect at least 300 pg/mL of recombinant human MCP-1 when used with our biotinylated antigen affinity purified anti-human MCP-1 (XP-5218Bt) as the detection antibody at a concentration of approximately 0.5-1.0 ug/mL. Western Blot: To detect hMCP-1 by Western Blot analysis this antibody can be used at a concentration of 0.20-0.40 ug/mL. Used in conjunction with compatible secondary reagents the detection limit for recombinant hMCP-1 is 1.0-2.0 ng/lane, under reducing or non-reducing conditions.
Type: Primary
Antigen: CCL2
Clonality: Monoclonal
Clone: 2.2-4H5-01A11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human