Specifications
- Antibody Type:Primary
- Antigen Name:chemokine (C-C motif) ligand 8
- Antigen Symbol:CCL8
- Clonality:Monoclonal
- Clone:1.1_2D4-1A3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western Blot:Yes
- Size:0.5 mg
- Neutralisation:Yes
- Form:Lyophilized
- Gene ID:P80075
- Antigen Synonyms:CCL8
- Storage Temperature:Stable at room temperature for at least one month and for greater than a year when kept at –20 °C. Reconstitute in sterile water to a concentration of 0.1 - 1.0 mg/mL. Stable for at least six weeks at 2-4 °C when reconstituted.
- Shipping Temperature:RT
- Immunogen:The monoclonal antibody was derived form BALB/c X ICR F1 mice and belongs to the IgG1K isotype.
- Purification:The monoclonal antibody against human MCP-2 designated as VL-1 was purified from ascites fluid on a Protein A affinity column.
- Cat. No.:10813-678
- Supplier no.:XP-5730-M
Specifications
About this item
MCP 2 is a chemotactic factor that attracts monocytes, lymphocytes, basophils and eosinophils. MCP 2 is important to inflammatory host responses, and is found in the highest concentration in the small intestine and peripheral blood cells.
Neutralization: To yield one-half maximal inhibition [ND50] of the biological activity of hMCP-2 (100 ng/mL), a concentration of 0.8-1.0 ug/mL of this antibody is required. ELISA: In a sandwich ELISA (assuming 100 uL/well), a concentration of 1.0-2.0 ug/mL of this antibody will detect at least 100 pg/mL of recombinant human MCP-2 when used with our biotinylated antigen affinity purified anti-human MCP-2 (XP-5224Bt) as the detection antibody at a concentration of approximately 0.5-1.0 ug/mL. Western Blot: To detect hMCP-2 by Western Blot analysis this antibody can be used at a concentration of 0.25-0.50 ug/mL. Used in conjunction with compatible secondary reagents the detection limit for recombinant hMCP-2 is 0.25-0.50 ng/lane, under non-reducing conditions.
Type: Primary
Antigen: CCL8
Clonality: Monoclonal
Clone: 1.1_2D4-1A3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human